Learn More
Enterprise Technology Solutions Paint Marker, Fine Bullet Tip, White, Dozen
Supplier: Enterprise Technology Solutions NSN2074159
ABILITYONE 7520012074159 Skilcraft Pump-action, fiber tip, oil-based paint markers for use on porous and non-porous surfaces including stone, wood, glass, plastic, metal, leather, ceramic, marble, granite, glossy surfaces, paper and more. Ideal for heavy-duty industrial and professional applications as well as for personalization and decorative purposes. Quick-drying paint resists water, sunlight, rough handling and various chemicals. It's weather-proof, fade-proof and acid-free making it archival safe. Ergonomic rubber grip for added comfort and a durable pocket clip on the cap.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.