Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ 7500PF Nitrile Disposable Glove
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Description
- Engineered with SHOWA's revolutionary Eco Best Technology™ (EBT), the SHOWA 7500PF disposable glove is biodegradable, achieving 82% biodegradation in 386 days in a laboratory.
- A rolled cuff prevents dirt from entering, and the powder and latex-free design reduces risk of discomfort and allergies.
- Textured surface grip makes wet work safer and easier.
Specifications
Certifications/Compliance | ISO 374-1:2016, ISO 374-5:2016 |
Applications | Laboratory |
Color | Blue |
Disposable | Single-use |
Exterior Finish | Fully Textured |
Finger Thickness (English) | 4 mil |
Latex Free | Yes |
Material | Nitrile |
Powdered | Powder Free |
Size | X-Small |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title