Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ 75 cm Surgical Suture Thread Variety Pack
SCI_ED

This mixed variety pack of surgical suture threads includes nylon, silk, polyester, and polypropylene threads with curved needle.

Supplier:  Walter Products™ SNM12

Catalog No. S139045


Add to cart

Alert:

Not intended for use on human or animal subjects.


Description

Description

  • Mixed variety of 75 cm surgical suture threads
  • Superb strength that will not fatigue with normal flexing
  • Exceptionally smooth surface, superior tensile strength, and excellent knot security
  • Ultra sharp, 3/8 reverse cutting, stainless steel needle
  • Sterile and ready to use
  • Excellent for suture training and practice
Specifications

Specifications

Suturing Practice
6 Pk. Nylon, 6 Pk. Silk, 6 Pk. Polyester, 6 Pk. Polypropylene Thread with Needle
18 mm
4-0
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit