Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ 7166 Nitrile Coated Oil & Grease Resistant Gloves
Click to view available options
Exterior Finish:
Fully Textured
No Texture
Description
- Exclusive coating provides cut, puncture, snag, and abrasion resistance
- Smooth finish enhances grip in dry and oily conditions
- Rough coating enhances grip and adds durability
- Barrier is resistant to oil and grease, and prevents oil breakthrough
- Reinforced safety cuff for additional protection
- Economical replacement for leather
Specifications
Construction | Cut & Sewn |
Cuff Material | Reinforced Cotton |
Coating Material | Nitrile |
Color | Black |
Exterior Finish | No Texture |
Hand | Specific |
Material | Cotton |
Product Type | Dip-Coated Gloves |
Size | Large |
For Use With (Application) | Chemical Spray/Treatment, Plumbing, Landscaping, and Utility Work |
Show More |