Learn More
Enterprise Technology Solutions 7045015584993, Skilcraft Usb Flash Drive With 256-Bit Aes Encryption, 8 Gb, Blue
Supplier: Enterprise Technology Solutions NSN5584993
Portable storage device with 256-bit AES Encryption. FIPS 140-2, Level 1 Certified, Certificate No. 347 ISC. Protection against brute force attacks. Preloaded UltraLockâ„¢ software ready for password customization. No back door entry for maximum security. Durable and reliable storage data retention for 10+ years.