Learn More
Enterprise Technology Solutions APC Essential SurgeArrest 6-Outlet Surge Protector (6', Black, 120V
SDP
Supplier:  Enterprise Technology Solutions PE66
Encompass
The Essential SurgeArrest 6-Outlet Surge Protector from APC provides you with six outlets for you to connect electrical devices such as computers, TVs, and more. It protects your connected equipment from electrical power surges and spikes, and also features EMI and RFI noise filtering, which reduces line noise that can cause data loss. This surge protector has a lifetime warranty and a $50,000 connected equipment warranty. Furthermore, it features Fail Safe Mode, which disconnects your equipment from the power supply should it be compromised, ensuring your devices aren't damaged.
Catalog No. 50-209-7237