Learn More
Enterprise Technology Solutions APC 6-Outlet SurgeArrest Surge Protector (Black, 15' Cable
SDP
Supplier:  Enterprise Technology Solutions PE615
Encompass
Squash surges and more with the black 6-Outlet SurgeArrest Surge Protector from APC. Featuring a NEMA 5-15P input connection and six NEMA 5-15R outlets, this surge protector has a surge energy rating of 1080 joules. In addition to EMI and RFI filtering, users also benefit from a fail safe mode with working and wiring fault indicators. This surge protector is capable of an output up to 120 VAC at 50/60 Hz and 15A. Its power cable measures 15'.
Catalog No. 50-209-7242