Learn More
METTLER TOLEDO DO Sensor InPro6850i/12/220
Supplier: METTLER TOLEDO 52206120
The InPro 6850i dissolved oxygen sensor with 12mm diameter body provides maximum accuracy and ultimate cleanability for large and small vessels with limited space or containers with smaller volumes.