Learn More
Nelson Jameson Company 3M PETRIFILM Yeast and Mold Count Plates
Supplier:  Nelson Jameson Company 4603407 C
Encompass
Yeast and Mold Count Plates determine air and food borne yeasts and molds which can invade dry ingredients, processing equipment and storage facilities. Maximize productivity by increasing your laboratory efficiency with 3M™ Petrifilm™ Plates. SPREADER INCLUDED. These sample-ready plates streamline your testing process, reducing the labor hours needed to perform microbial testing. Fast. Efficient. And there's a full line of Petrifilm Plates so you can monitor a variety of conditions for quality. AOAC approved.
Catalog No. NC1797199