Learn More
Purus™ 60-Layer Cleanroom Tacky Mats: PROMO
Remove dirt and dust from foot traffic and equipment wheels
Supplier: Purus™ PS 3645 64 GPM
Description
- 1.4mil low-density polyethylene film layers fully coated with a particle grabbing acrylic adhesive
- Suitable for use in industrial, semiconductor, pharmaceutical, biotech and telecommunications cleanrooms
- Consistent adhesive coverage for maximum performance on each layer
- Numbered tabs ensure one-sheet-at-a-time removal and indicate number of remaining layers
Specifications
91.4 x 114.3 cm (36 x 45 in.) | |
Gray | |
60 Sheets/Mat |
Suitable for use in industrial, semiconductor, pharmaceutical, biotech, and telecommunications cleanrooms | |
Polyethylene |