Learn More
Purus™ 60-Layer Cleanroom Tacky Mats: PROMO
Remove dirt and dust from foot traffic and equipment wheels
Supplier:  Purus™ PS 1845 64 BPM
 View more versions of this product
Catalog No. 19-125-816PM
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
View alternate product
Description
Description
  • 1.4mil low-density polyethylene film layers fully coated with a particle grabbing acrylic adhesive
  • Suitable for use in industrial, semiconductor, pharmaceutical, biotech and telecommunications cleanrooms
  • Consistent adhesive coverage for maximum performance on each layer
  • Numbered tabs ensure one-sheet-at-a-time removal and indicate number of remaining layers
Specifications
Specifications
45.7 x 114.3 cm (18 x 45 in.)
60 Sheets/Mat
Polyethylene
Suitable for use in industrial, semiconductor, pharmaceutical, biotech, and telecommunications cleanrooms
Blue
Product Suggestions
Product Suggestions