Learn More
Grainger 6' SLF REG HEATING CABLE 120VA
Supplier:  Grainger 13R101
Encompass_Preferred
6 ft. Self Regulating Heating Cable, Wet or Dry, Max. Circuit Length 200 ft., 120VAC
Catalog No. NC1773486