Learn More
Medchemexpress LLC HY-B0236 50mg Medchemexpress, 6-Aminocaproic acid CAS:60-32-2 Purity:>98%
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Medchemexpress LLC HYB023650MG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Medchemexpress, HY-B0236 50mg 6-Aminocaproic acid CAS:60-32-2 6-Aminocaproic acid (EACA), a monoamino carboxylic acid, is a potent and orally active inhibitor of plasmin and plasminogen. 6-Aminocaproic acid is a potent antifibrinolytic agent. 6-Aminocaproic acid prevents clot lysis through the competitive binding of lysine residues on plasminogen, inhibiting plasmin formation and reducing fibrinolysis. 6-Aminocaproic acid can be used for the research of bleeding disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.