Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone 98.0+%, TCI America™
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 196°C |
Quantity | 5 g |
Formula Weight | 203.25 |
Percent Purity | ≥98.0% (GC,T) |
Physical Form | Crystalline Lumps |
Chemical Name or Material | 6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone |
Safety and Handling
TSCA : No