Learn More
Perkin Elmer US LLC 5X Beam Condenser
Supplier: Perkin Elmer US LLC 01860794
Benefits of the 5X Beam Condenser include: Easy measurement of microgram and submicrogram quantities of sample Highly efficient optical design results in high-quality IR spectra Miniature Diamond Anvil CellTM; Available for high-pressure analysis of small, optically thick polymeric samples. An Optional fiber holder used for single fiber analysis.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.