Learn More
Zymo Research Corporation 5-Methylcytosine & 5-HydroxymethylcytosineDNA Standard Set, full set
Supplier: Zymo Research Corporation D5405
The only difference is that each contains either 100% unmodified cytosines, 5-methylcytosines, or 5-hydroxymethylcytosines. Since the sequence and extent of cytosine modification is known, this DNA standard set is ideal for use in calibration of various applications intended for quantitation of cytosine modifications.