Learn More
Chemglass Life Sciences 50mL Glassblowers Pear Recovery Blank
Supplier: Chemglass Life Sciences CG62203
50mL Glassblowers Pear Recovery Blank. Blanks for fabrication of recovery flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.