Learn More
Chemglass Life Sciences 50mL Ampul, Drying, Airfree, Schlenk, Round Bottom, 35 O.D. x 200mm Height
Supplier: Chemglass Life Sciences AF008404
50mL Ampul, Drying, Airfree, Schlenk, Round Bottom, 35 O.D. x 200mm Height. Round bottom, cylindrical drying ampuls with 10mm OD top stem. Manufactured from Type 1 borosilicate glass. Please note: Ampuls are available with top stems to fit AF-0067 Cajon Unions.