Learn More
Teledyne ISCO, Inc Silicone Rubber Pump Tubing / Discharge Tubing Bulk Roll 50 ft
Supplier: Teledyne ISCO, Inc 606700047
Used as pump tubing on the GLS, Glacier, and 3700 Series Samplers. Used as discharge tubing on the Avalanche, GLS, Glacier, 4700, 5800 and 6700 Series Samplers. Dimensions: 0.375 Inch I.D. x 0.625 Inch O.D. x 0.125 Inch Wall.