Learn More
ORS Nasco CART OV/AG P100 HEPA 2/PK
Supplier: ORS Nasco 0687583P100L
Honeywell part# 7583P100L. Honeywell North Low-Profile Cartridges and Cartridge/P100 Filter Combinations Cartridge Type: Filter Cartridge Yellow Magenta Quantity: 12 Case Applications: Industrial applications For Use With: Respirator Organic Vapor/Acid Gas Assembly Certifications/Compliance: NIOSH 42CFR84