Learn More
ORS Nasco PORTA STREAM II 16 GALLON
Supplier: ORS Nasco 203320002000000
Honeywell part# 32-000200-0000. Honeywell Fendall Porta Stream II Eyewash Station Capacity: 60.5L 2048 oz. 60566.58mL 16 gal. Includes: 180 oz. Eyesaline concentrate Certifications/Compliance: ANSI Z358.1 - Meets ANSI Z358.1 15-minute flushing requrement Hi viz green Depth: 19 in. 48.26cm Flow Rate: 0.4 gal./min.