Learn More
Trajan Scientific SYR AUTO SK 5F HP 0.47 6/PK
Supplier: Trajan Scientific 001804
SGE Autosampler Syringes with 42 mm Cone Tip Fixed Needle, 5 uL, Description: Not gas tight, Diameter Outer Needle: 0.47 mm, Needle Gauge: 26 G, Connection Type: Fixed, For Use With: Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS, Gas chromatography, Length Needle: 42 mm, Volume: 5 uL