Learn More
Trajan Scientific SK 10F-AG-GT-0.63C SYRNGE 25PK
Supplier: Trajan Scientific 002816
Trajan Scientific and Medical Autosampler Syringes for Agilent GCs, Fixed Needle, Volume: 10 uL, Needle Gauge: 23 G, Diameter Outer Needle: 0.63 mm, Description: Agilent Syringe, Needle Point Style: Cone