Learn More
Trajan Scientific SYRINGE SHIMADZU 10R-S-0.63
Supplier: Trajan Scientific 002898
Trajan Autosampler Syringes, Connection Type: Removable, Volume: 10 uL, For Use With: Shimadzu Models AOC14, AOC17, AOC20, AOC20i, Gas chromatography, Needle Gauge: 23 G, Length Needle: 42 mm, 1.6 in., Diameter Outer Needle: 0.02 in., 0.63 mm, Length: 1.65 in., 42 mm, Model: 10R-S-0.63C