Learn More
Trajan Scientific SYR 10UL 10R-C/F-5/0.47C 6/PK
Supplier: Trajan Scientific 002986
SGE PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle, Description: Not gas tight, Volume: 10 uL, Diameter Outer Needle: 0.47 mm, Needle Gauge: 26 G, Length Needle: 50 mm, Connection Type: Fixed