Learn More
Trajan Scientific DIAMOND MS SYRINGE
Supplier: Trajan Scientific 003790
Trajan Diamond MS Thermo RSH and CTC RTC Autosampler Syringes, Volume: 25 uL, Model: 25R-RTC/RSH-GT-MS1, Connection Type: Removable, For Use With: Gas/Liquid chromatography, Thermo RSH and CTC RTC, Max. Temperature: 662 deg.F, 350 deg.C, Quantity: Each