Learn More
Trajan Scientific SYR 25UL 25F-C/F-GT-5/.63C
Supplier: Trajan Scientific 003987
SGE PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle, Description: Gas tight, Volume: 25 uL, Diameter Outer Needle: 0.63 mm, 0.024 in., Needle Gauge: 23 G, Length Needle: 50 mm, 1.96 in., Connection Type: Fixed, Diameter Inner Needle: 0.009 in., 0.24 mm, Diameter Outer: 6.5 mm