Learn More
Trajan Scientific 100F-C/F-GT-5/0.63C
Supplier: Trajan Scientific 005335
SGE PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle, Description: Gas tight, Volume: 100 uL, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Length Needle: 50 mm, Connection Type: Fixed, For Use With: Gas chromatography, CTC, LEAP, CombiPAL, GC PAL, Model: 100F-C/T-GT-5/0.63C