Learn More
Trajan Scientific SYRINGE 5MR-LL-GT
Supplier: Trajan Scientific 008760
SGE Gastight Syringes: Removable Luer-Lok Models, Connection Type: Removable, Diameter Outer: 0.53 in., 13.7 mm, Model: 5MDR-LL-GT, Needle Gauge: No Needle Included, Volume: 5.0 mL, Accuracy: +/-1%, For Use With: Suitable for gases or liquids for maximum inertness