Learn More
Trajan Scientific SYRINGE 10MR-LL-GT
Supplier: Trajan Scientific 008960
SGE Gastight Syringes: Removable Luer-Lok Models, Connection Type: Fixed, Diameter Outer: 0.7 in., 18 mm, Model: 10MDR-LL-GT, Needle Gauge: No Needle Included, Volume: 10.0 mL, Accuracy: +/-1%, For Use With: Suitable for gases or liquids for maximum inertness