Learn More
Trajan Scientific SYR AUTO VAR 10UL 10R-VA8X-II
Supplier: Trajan Scientific 002924
Trajan Autosampler Syringes, Connection Type: Removable, Volume: 10 uL, For Use With: Varian 8035, 8100 and 8200, Gas chromatography, Needle Gauge: 25 G, Length Needle: 53 mm, 2.08 in., Diameter Outer Needle: 0.019 in., 0.5 mm, Length: 2 in., 53 mm, Model: 10R-GT-VA8X-2