Learn More
Trajan Scientific SYRINGE 25MDR-LL-GT
Supplier: Trajan Scientific 009462
SGE Gastight Syringes: Removable Luer-Lok Models, Connection Type: Removable, Diameter Outer: 0.31 in., 8 mm, Model: 25MDR-LL-GT, Needle Gauge: No Needle Included, Volume: 25.0 mL, Accuracy: +/-1%, For Use With: Suitable for gases or liquids for maximum inertness