Learn More
Trajan Scientific CPLRY 30QC2.5/BPX5-1.00
Supplier: Trajan Scientific 054122
Trajan BPX5 GC Capillary Columns, Diameter Inner: 0.25 mm, Film Thickness: 1 um, Length: 30 m, For Use With: Solvents Analysis, Pesticides and Herbicides Analysis, Hydrocarbons, Phenols, Amines, GC/MS, Packing Material: BPX5, Stationary Phase: 5% Phenyl Polysilphenylene Siloxane