Learn More
Strategic Applications Inc 500ul K3 EDTA Blood Tube, Easy Cap (Purple Top), 100/pk
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Strategic Applications Inc MVE500
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
This item has a minimum order quantity of 10 packs per supplier requirements. 500uL K3EDTA sample tube with collection rim. Twist-off, push-on cap. Slim profile ensures fit in standard racks and centrifuges. 100/box. Product is for laboratory animal research only. ** If a COA is required it must be requested when the order is placed or a $50.00 fee will be apply. Contact Thermo Fisher Customer Service at 800-766-7000 to request to COA. **
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.