Learn More
Ace Glass, Inc. Master Heat Gun; Model HG-501D, 1200°F inside the nozzle, 552°F one inch from the nozzle, cULus rated
Supplier: Ace Glass, Inc. 1207312
Master Heat Gun; Model HG-501D, 1200°F inside the nozzle, 552°F one inch from the nozzle, cULus rated