Learn More
Invivogen 5' triphosphate double stranded RNA (5' ppp-dsRNA)
Supplier: Invivogen TLRL3PRNA100
5’ triphosphate double stranded RNA (5’ ppp-dsRNA) is a synthetic ligand for retinoic acid-inducible protein I (RIG-I). RIG-I is a cytosolic pattern recognition receptor that senses pathogen-associated molecular patterns (PAMPs) on viral RNA and triggers an antiviral immune response by the activation of type-I interferons (IFN-α and -β).