Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Phenyl-1-pentenylboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog No. AAL19701MD
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154820-97-0
Molecular Formula C17H25BO2
Molecular Weight (g/mol) 272.195
MDL Number MFCD03788752
InChI Key HLUHQMVNJDKHRO-GXDHUFHOSA-N
Synonym 5-phenyl-1-pentenylboronic acid pinacol ester, e-4,4,5,5-tetramethyl-2-5-phenylpent-1-en-1-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1e-5-phenylpent-1-en-1-yl-1,3,2-dioxaborolane
PubChem CID 10945575
IUPAC Name 4,4,5,5-tetramethyl-2-[(E)-5-phenylpent-1-enyl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C=CCCCC2=CC=CC=C2

Specifications

Refractive Index 1.498
Quantity 250 mg
Beilstein 7295229
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 272.2
Percent Purity 96%
Chemical Name or Material 5-Phenyl-1-pentenylboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.