Learn More
US Plastic Corporation 5 Gallon Heavyweight Natural HDPE Carboy
Supplier: US Plastic Corporation 75001
The Fort-Pak is a rigid plastic drum molded of high density polyethylene featuring a 2" closure and a center 3/4" NPT reducer in the cap. These UN rated carboys are ideal for shipping regulated liquids.