Learn More
Med Vet International Ethicon Ethilon Nylon Suture, Size 5-0, P-3, 18", 12/Box
Supplier: Med Vet International 698GRL
The Ethicon 698G suture is a high-quality non-absorbable Nylon surgical suture. It measures 18in. in length, has a size of 5-0, and are dyed black to provide greater visibility while suturing. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.