Learn More
Wilmad Labglass 4MM THIN EPR TUBE QUARTZ
Supplier: Wilmad Labglass 707SQ100M
Precision brand EPR tubes hold 10-15 times tighter tolerance on the inner diameter, and >5 times smaller camber and concentricity over Economy Brand tube. Tube Length 100mm. Tube OD 4mm. Tube ID 3mm.