Learn More
Revvity Health Sciences Inc 4 mL MicroBeta Vial, 3000/pk
Supplier:  Revvity Health Sciences Inc 1200421
Encompass
Revvity offers high quality glass and plastic vials for liquid scintillation counting. The best vial to choose is dependent on the type and volume of sample to be counted and the cocktail that will be used. Why Choose Plastic? Lower background levels versus glass vials Higher counting efficiency versus glass vials Combustible - easier waste disposal No solvent permeation with safer, high-flash point cocktails
Catalog No. 50-905-1928