Learn More
Cole-Parmer Solid PP Cable/Zip Ties, 4" L, White; 1000/Pk
Supplier: Cole-Parmer 0683085
Polypropylene (PP) cable ties are ideal for tubing bundles. PP resists a number of chemicals and is UV-resistant.