Learn More
Seracare Life Sciences Inc 4CN 2-C MEMB HRP SUBSTRATE
Supplier:  Seracare Life Sciences Inc 54200020
Encompass_Preferred
An ideal alternative to other, more sensitive substrates where background is a problem. 4 CN (4-chloro-1-napthol) produces a precipitate at the reaction site in the presence of peroxidase-labeled conjugates. It is optimized for use in membrane assays and dot blots. 2-component format, simply mix and use.
Catalog No. 50-674-18