Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cellucap 1.25 oz. Spunbound Polypropylene Coveralls
Non-woven material offers lightweight splash protection from non-hazardous liquids and dry particulates
$175.00 - $203.00
Specifications
Material | Spunbond Polypropylene |
---|---|
Color | White |
Description | 1.25 oz. Spunbound Polypropylene |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-133-906
|
Cellucap
55840XXXX |
4X-Large |
Case of 25 for $203.00
|
|
19-133-905
|
Cellucap
55840XXX |
3X-Large |
Case of 25 for $193.00
|
|
19-133-902
|
Cellucap
55840L |
Large |
Case of 25 for $175.00
|
|
19-133-900
|
Cellucap
55840S |
Small |
Case of 25 for $175.00
|
|
19-133-901
|
Cellucap
55840M |
Medium |
Case of 25 for $175.00
|
|
Specifications
Spunbond Polypropylene | |
White | |
1.25 oz. Spunbound Polypropylene |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title