Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Novagen™ HT96™ NovaBlue Competent Cells
High-efficiency competent cells predispensed in 96-well plate for high-throughput applications
$756.00 - $2840.00
Specifications
Efficiency | > 1 x 108CFU/μg |
---|---|
Content And Storage | Store HT96 Competent Cells, SOC Medium and Test Plasmid at -70°C or below. Store Cap Strips, Plate Lids and Reagent Reservoirs at room temperature |
For Use With (Application) | For high-throughput transformation |
Includes: HT96 NovaBlue Competent Cells (1 or 4 plates), SOC Medium (1 × 14mL or 4 × 14mL), Test Plasmid (1 × 10μL or 2 × 10 μL), 8-Cap Strips (1 or 4), Reagent Reservoirs (1 or 4), Plate Lids (1 or 4)
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
71-011-3MI
|
MilliporeSigma™
710113 |
1 Plate |
Each for $756.00
|
|
|||||
71-011-4MI
|
MilliporeSigma™
710114 |
4 Plates |
Each for $2,840.00
|
|
|||||
Description
NovaBlue is a K-12 strain ideally suited as an initial cloning host because it has high transformation efficiency, blue/white screening capability (with appropriate plasmids) and mutations in recA and endA which result in high yields of high-quality plasmid DNA.
- Cells dispensed in 20μL volumes in 96 well polypropylene plate compatible with thermal cyclers and water baths
- Wells are individually sealed and have raised rims to prevent cross-contamination
- Seals can be easily pierced with standard pipet tips or removed for access
- Strips of caps are also provided for reliable sealing during manipulation and storage
- Groups of 24 wells can be easily split from the plate for processing smaller numbers of samples
- Efficiency: >1 × 108CFU/μg
Specifications
> 1 x 108CFU/μg | |
Store HT96 Competent Cells, SOC Medium and Test Plasmid at -70°C or below. Store Cap Strips, Plate Lids and Reagent Reservoirs at room temperature | |
For high-throughput transformation |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title