Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Graduated Cylinders, PP, Round Base

Round Base provides good stability.
Supplier: Eisco™ CH0353A
This item is not returnable.
View return policy
Description
- Made of polypropylene
- Raised graduations
Specifications
10 mL | |
1 Each | |
Autoclavable | |
Round |
Polypropylene | |
0.2 mL | |
Round | |
Graduated Measuring Cylinder |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title