Learn More
Brady™ MRO Plus™ Absorbent Rolls
Strong, durable and low linting, making them the perfect go-to products for nearly every industrial setting
Supplier: Brady™ MRO30P
Description
- Highly versatile, gray absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
- Heavy weight - specifically designed for continuous leaks and larger spills so you use less material
- Medium weight - optimal for most general maintenance needs
- 3-ply Meltblown Polypropylene construction is designed for general industrial applications
- Non-perforated absorbents offer the greatest absorbency and better tear resistance in high traffic areas
- Perforations provide cost control and reduce waste by allowing you to use only what you need