Learn More
MilliporeSigma Omnipore Membrane Filter, 100/pk
Supplier:  MilliporeSigma JAWP04700
Encompass
Omnipore Membrane Filter, PTFE, Hydrophilic, 1.0um, 47mm, Plain Surface, White, Screen filter type, 130deg.C, Porosity %: 80
Catalog No. NC9296429