Learn More
HANNA INSTRUMENTS INC 470 mV @ 25°C ORP Test Solution (500 mL)
SDP
Supplier:  HANNA INSTRUMENTS INC HI7022L
Encompass
HI7022L is a quality ORP test solution made from reagent grade chemicals and used to test platinum and gold ORP electrodes. Hanna test solutions have the lot number and expiration date clearly marked on the label and are air tight with a tamper-proof seal to ensure the quality of the solution.Prepared with reagent grade chemicalsAllows for a quick check of your ORP electrodeAir-tight seal to ensure the quality of the solution
Catalog No. NC1675730