Learn More
Electron Microscopy Sciences Multi Pin Holder 45 degree for 3 pin stubs
Supplier: Electron Microscopy Sciences 7595260
This 25.4 mm in diameter 45 Degree multi pin holder is designed to save time. It allows for a higher SE signal without having to tilt the specimen It is made from machined aluminum with stainless steel Allen set screws. It accommodates up to three standard 12.7 mm (1/2") pin stubs with 3.2 mm (1/8") pin.